4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone 97%

Lieferant: Apollo Scientific

PC8140-1G PC8140-500MG PC8140-5G
APOSPC8140-1GEA 533 EUR
APOSPC8140-1G APOSPC8140-500MG APOSPC8140-5G
4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone 97%
4-Chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-piperazino-3(2H)-pyridazinone
Formel: C₁₄H₁₂Cl₂F₃N₅O
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793846
CAS-Nummer: 338399-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR