4-Chlor-2-fluorbenzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC8145-25G PC8145-5G
APOSPC8145-25GEA 43.5 EUR
APOSPC8145-25G APOSPC8145-5G
4-Chlor-2-fluorbenzylalkohol 98%
4-Chlor-2-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143285
CAS-Nummer: 56456-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR