4-Chlor-3-fluorzimtsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC8154-10G
PC8154-1G
PC8154-5G
PC8154-25G
PC8154-100G
APOSPC8154-10GEA
136
EUR
APOSPC8154-10G
APOSPC8154-1G
APOSPC8154-5G
APOSPC8154-25G
APOSPC8154-100G
4-Chlor-3-fluorzimtsäure
4-Chlor-3-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Molecular Weight: 200.6 g/mol Schmelzpunkt: 241…245 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143294 CAS-Nummer: 202982-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...