1-(3-Chloro-4-fluorophenyl)propan-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Chloro-4-fluorophenyl)-1-propanone, 3'-Chloro-4'-fluoropropiophenone
PC8168-5G
PC8168-1G
APOSPC8168-5GEA
166
EUR
APOSPC8168-5G
APOSPC8168-1G
1-(3-Chloro-4-fluorophenyl)propan-1-one
1-(3-Chloro-4-fluorophenyl)propan-1-one
Formel:
C₉H₈ClFO Molecular Weight: 186,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143013 CAS-Nummer: 194943-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...