1-Chlor-2,3,4-trifluorbenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8169-25G
PC8169-5G
APOSPC8169-25GEA
65.4
EUR
APOSPC8169-25G
APOSPC8169-5G
1-Chlor-2,3,4-trifluorbenzol 97%
1-Chloro-2,3,4-trifluorobenzene
Formel:
C₆H₂ClF₃ Molecular Weight: 166,5300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042222 CAS-Nummer: 36556-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...