2,6-Dibrom-4-fluoranisol

Lieferant: Apollo Scientific
Achtung

PC8172-25G PC8172-5G
APOSPC8172-25GEA 302 EUR
APOSPC8172-25G APOSPC8172-5G
2,6-Dibrom-4-fluoranisol
2,6-Dibrom-4-fluoranisol
Formel: C₇H₅Br₂FO
Molecular Weight: 283.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042231
CAS-Nummer: 443-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR