4-Fluor-o-anisaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluor-2-anisaldehyd, 4-Fluor-2-methoxybenzaldehyd

PC8206-1G PC8206-25G PC8206-5G
APOSPC8206-1GEA 21.9 EUR
APOSPC8206-1G APOSPC8206-25G APOSPC8206-5G
4-Fluor-o-anisaldehyd 98%
4-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
Schmelzpunkt: 57…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143318
CAS-Nummer: 450-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR