2-Fluorthiobenzamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluorthiobenzoesäureamid

PC8237-1G PC8237-25G PC8237-5G
APOSPC8237-1GEA 84.6 EUR
APOSPC8237-1G APOSPC8237-25G APOSPC8237-5G
2-Fluorthiobenzamid
2-Fluorthiobenzamid
Formel: C₇H₆FNS
Molecular Weight: 155.2 g/mol
Schmelzpunkt: 83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04973316
CAS-Nummer: 75907-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR