4-Chlor-2,6-difluorbenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC8238-1G PC8238-25G PC8238-5G
APOSPC8238-1GEA 21.9 EUR
APOSPC8238-1G APOSPC8238-25G APOSPC8238-5G
4-Chlor-2,6-difluorbenzaldehyd 98%
4-Chloro-2,6-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
Schmelzpunkt: 68…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094460
CAS-Nummer: 252004-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR