Methyl-4-trifluoromethylbenzoylacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionoate, 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionic acid methyll ester, Methyl 4-(Trifluoromethyl)benzoylacetate, 4-(Trifluoromethyl)benzoylacetic acid methyll ester, Methyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate

PC8256-1G PC8256-5G
APOSPC8256-1GEA 21.9 EUR
APOSPC8256-1G APOSPC8256-5G
Methyl-4-trifluoromethylbenzoylacetate
Methyl-4-trifluoromethylbenzoylacetate
Formel: C₁₁H₉F₃O₃
Molecular Weight: 246,185 g/mol
Schmelzpunkt: 57…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216521
CAS-Nummer: 212755-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR