3,4,5-Trifluorzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4,5-Trifluorcinnamsäure, 3-(3,4,5-Trifluorphenyl)propensäure

PC8270-25G PC8270-5G
APOSPC8270-25GEA 168 EUR
APOSPC8270-25G APOSPC8270-5G
3,4,5-Trifluorzimtsäure 98%
3,4,5-Trifluorzimtsäure
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
Schmelzpunkt: 195…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083529
CAS-Nummer: 152152-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR