2,6-Difluor-4-hydroxybenzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8321-1G
PC8321-5G
APOSPC8321-1GEA
87
EUR
APOSPC8321-1G
APOSPC8321-5G
2,6-Difluor-4-hydroxybenzoesäure 97%
2,6-Difluoro-4-hydroxybenzoic acid
Formel:
C₇H₄F₂O₃ Molecular Weight: 174.1 g/mol Schmelzpunkt: 170…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094500 CAS-Nummer: 214917-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...