5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(3,4,4-trifluorobut-3-en-1-ylthio)-1,3,4-thiadiazole

PC8328-1G PC8328-250MG PC8328-5G
APOSPC8328-1GEA 173 EUR
APOSPC8328-1G APOSPC8328-250MG APOSPC8328-5G
5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine
5-[(3,4,4-Trifluoro-3-butenyl)sulphanyl]-1,3,4-thiadiazol-2-amine
Formel: C₆H₆F₃N₃S₂
Molecular Weight: 241,26 g/mol
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01443829
CAS-Nummer: 145070-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR