2-Brom-4-fluorthioanisol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-4-fluorophenyl)(methyl)sulphane

PC8331-1G PC8331-250MG PC8331-5G
APOSPC8331-1GEA 97.3 EUR
APOSPC8331-1G APOSPC8331-250MG APOSPC8331-5G
2-Brom-4-fluorthioanisol 96%
2-Brom-4-fluorthioanisol
Formel: C₇H₆BrFS
Molecular Weight: 221,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801042
CAS-Nummer: 91524-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR