4-Brom-2-fluorphenylessigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Bromo-2-fluorophenyl)acetic acid

PC8343-25G
APOSPC8343-25GEA 65.4 EUR
APOSPC8343-25G
4-Brom-2-fluorphenylessigsäure
4-Brom-2-fluorphenylessigsäure
Formel: C₈H₆BrFO₂
Molecular Weight: 233.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032952
CAS-Nummer: 114897-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR