O-[2-Oxo-1(2H)-pyridyl]-N,N,N′,N′-tetramethyluronium-tetrafluoroborat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: O-(2-Oxo-1(2H)pyridyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TPTU

PC8352-25G PC8352-5G PC8352-100G
APOSPC8352-25GEA 52.2 EUR
APOSPC8352-25G APOSPC8352-5G APOSPC8352-100G
O-[2-Oxo-1(2H)-pyridyl]-N,N,N′,N′-tetramethyluronium-tetrafluoroborat 98%
O-[2-Oxo-1(2H)-pyridyl]-N,N,N′,N′-tetramethyluronium-tetrafluoroborat
Formel: C₁₀H₁₆BF₄N₃O₂
Molecular Weight: 297,06 g/mol
Schmelzpunkt: 140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00075475
CAS-Nummer: 125700-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR