3-Fluor-D-phenylalanin
Lieferant: Apollo Scientific
Synonyme:
H-D-Phe(3-F)-OH
PC8359-1G
PC8359-25G
PC8359-5G
APOSPC8359-1GEA
234
EUR
APOSPC8359-1G
APOSPC8359-25G
APOSPC8359-5G
3-Fluor-D-phenylalanin
3-Fluor-D-phenylalanin
Formel:
C₉H₁₀FNO₂ Molecular Weight: 183,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066449 CAS-Nummer: 110117-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...