3-Chlor-5-fluorbenzylalkohol

Lieferant: Apollo Scientific
Achtung

PC8371-25G PC8371-5G PC8371-100G
APOSPC8371-25GEA 119 EUR
APOSPC8371-25G APOSPC8371-5G APOSPC8371-100G
3-Chlor-5-fluorbenzylalkohol
3-Chlor-5-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788554
CAS-Nummer: 79944-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR