2-Brom-2'-fluor-3'-(trifluormethyl)acetophenon
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Fluoro-3-(trifluoromethyl)phenacyl bromide
PC8405-1G
PC8405-25G
PC8405-5G
APOSPC8405-1GEA
58
EUR
APOSPC8405-1G
APOSPC8405-25G
APOSPC8405-5G
2-Brom-2'-fluor-3'-(trifluormethyl)acetophenon
2-Brom-2'-fluor-3'-(trifluormethyl)acetophenon
Formel:
C₉H₅BrF₄O Schmelzpunkt: 46,9…47.7 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094504 CAS-Nummer: 537050-11-6 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...