2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-piperazino}acetohydrazide 97%

Lieferant: Apollo Scientific

PC8420-1G PC8420-500MG PC8420-5G
APOSPC8420-1GEA 799 EUR
APOSPC8420-1G APOSPC8420-500MG APOSPC8420-5G
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-piperazino}acetohydrazide 97%
2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}acetohydrazide
Formel: C₁₂H₁₅ClF₃N₅O
Molecular Weight: 337.73 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141696
CAS-Nummer: 338979-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR