1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde 97%

Lieferant: Apollo Scientific

PC8428-1G PC8428-500MG
APOSPC8428-1GEA 379 EUR
APOSPC8428-1G APOSPC8428-500MG
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde 97%
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-carbaldehyde
Formel: C₁₁H₆ClF₃N₂O
Molecular Weight: 274.63 g/mol
Schmelzpunkt: 60…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231912
CAS-Nummer: 321430-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR