(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1-methyl-1H-imidazol-5-yl)methanol
Lieferant: Apollo Scientific
Synonyme:
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}-5-(hydroxymethyl)-1-methyl-1H-imidazole
PC8432-1G
PC8432-500MG
PC8432-5G
APOSPC8432-1GEA
533
EUR
APOSPC8432-1G
APOSPC8432-500MG
APOSPC8432-5G
(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1-methyl-1H-imidazol-5-yl)methanol
(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1-methyl-1H-imidazol-5-yl)methanol
Formel:
C₁₁H₉ClF₃N₃OS Molecular Weight: 323,73 g/mol Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00215151 CAS-Nummer: 338422-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...