3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1H-1,2,4-triazol-5-amine

Lieferant: Apollo Scientific

Synonyme: 5-Amino-3-[3-chloro-5-(trifluoromethyl)pyridin-2-ylthio]-1H-1,2,4-triazole

PC8435-1G PC8435-500MG PC8435-5G
APOSPC8435-1GEA 275 EUR
APOSPC8435-1G APOSPC8435-500MG APOSPC8435-5G
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1H-1,2,4-triazol-5-amine
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-1H-1,2,4-triazol-5-amine
Formel: C₈H₅ClF₃N₅S
Molecular Weight: 295.67 g/mol
Schmelzpunkt: 262…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00243559
CAS-Nummer: 250714-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR