2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Cyanophenyl)-2-(4-fluorophenyl)acetonitrile

PC8440-1G PC8440-500MG PC8440-5G
APOSPC8440-1GEA 172 EUR
APOSPC8440-1G APOSPC8440-500MG APOSPC8440-5G
2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile 97%
2-[Cyano(4-fluorophenyl)methyl]benzenecarbonitrile
Formel: C₁₅H₉FN₂
Molecular Weight: 236,25 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231953
CAS-Nummer: 116617-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR