2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Cyano-3-fluorophenyl)-2-(4-fluorophenyl)acetonitrile

PC8442-1G PC8442-500MG PC8442-5G
APOSPC8442-1GEA 172 EUR
APOSPC8442-1G APOSPC8442-500MG APOSPC8442-5G
2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile 97%
2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile
Formel: C₁₅H₈F₂N₂
Molecular Weight: 254,24 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231906
CAS-Nummer: 303150-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR