4-Chlor-3-fluorbenzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC8452-100G PC8452-25G PC8452-5G
APOSPC8452-100GEA 91.5 EUR
APOSPC8452-100G APOSPC8452-25G APOSPC8452-5G
4-Chlor-3-fluorbenzonitril 97%
4-Chlor-3-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155.56 g/mol
Schmelzpunkt: 79…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143291
CAS-Nummer: 110888-15-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR