2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC8456-1G
PC8456-250MG
APOSPC8456-1GEA
640
EUR
APOSPC8456-1G
APOSPC8456-250MG
2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
2-(2,6-Difluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
Formel:
C₁₀H₄F₂N₄O₂ Schmelzpunkt: 235…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00794280 CAS-Nummer: 338982-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...