3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile 97%

Lieferant: Apollo Scientific

PC8457-10G PC8457-1G PC8457-5G
APOSPC8457-10GEA 299 EUR
APOSPC8457-10G APOSPC8457-1G APOSPC8457-5G
3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile 97%
3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile
Formel: C₁₁H₁₁FN₂
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138582
CAS-Nummer: 320416-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR