2-(Dimethylamino)-6-(trifluoromethyl)-4-pyrimidinol

Lieferant: Apollo Scientific

Synonyme: 2-(Dimethylamino)-4-hydroxy-6-(trifluoromethyl)pyrimidine

PC8459-1G PC8459-5G
APOSPC8459-1GEA 93.1 EUR
APOSPC8459-1G APOSPC8459-5G
2-(Dimethylamino)-6-(trifluoromethyl)-4-pyrimidinol
2-(Dimethylamino)-6-(trifluoromethyl)-4-pyrimidinol
Formel: C₇H₈F₃N₃O
Molecular Weight: 207,16 g/mol
Schmelzpunkt: 176…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00171894
CAS-Nummer: 55545-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR