2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Ethylamino)-4-hydroxy-6-(trifluoromethyl)pyrimidine
PC8467-1G
PC8467-5G
APOSPC8467-1GEA
143
EUR
APOSPC8467-1G
APOSPC8467-5G
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol
2-(Ethylamino)-6-(trifluoromethyl)-4-pyrimidinol
Formel:
C₇H₈F₃N₃O Molecular Weight: 207,16 g/mol Schmelzpunkt: 198…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171969 CAS-Nummer: 338792-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...