2-Fluoro-4-iodonicotinaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-iodopyridine-3-carboxaldehyde
PC8525-1G
PC8525-5G
APOSPC8525-1GEA
21.9
EUR
APOSPC8525-1G
APOSPC8525-5G
2-Fluoro-4-iodonicotinaldehyde
2-Fluoro-4-iodonicotinaldehyde
Formel:
C₆H₃FINO Molecular Weight: 250,9970 g/mol Schmelzpunkt: 78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095290 CAS-Nummer: 153034-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...