trans-2-(4-Fluorophenyl)vinylboronic acid

Lieferant: Apollo Scientific
Achtung

PC8529-1G PC8529-5G
APOSPC8529-1GEA 158 EUR
APOSPC8529-1G APOSPC8529-5G
trans-2-(4-Fluorophenyl)vinylboronic acid
trans-2-(4-Fluorophenyl)vinylboronic acid
Formel: C₈H₈BFO₂
Molecular Weight: 165.96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093725
CAS-Nummer: 214907-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR