Pentafluoroethyl ethyl ether 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Ethyl perfluoroethyl ether
PC8543-25G
PC8543-5G
APOSPC8543-25GEA
1260
EUR
APOSPC8543-25G
APOSPC8543-5G
Pentafluoroethyl ethyl ether 98%
Pentafluoroethyl ethyl ether
Formel:
C₄H₅F₅O Molecular Weight: 164,08 g/mol Siedepunkt: 29…30 °C Dichte: 0,936 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862046 CAS-Nummer: 22052-81-9 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...