Methyl-5-fluor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
PC8595-1G
PC8595-5G
APOSPC8595-1GEA
183
EUR
APOSPC8595-1G
APOSPC8595-5G
Methyl-5-fluor-2-benzo[b]thiophencarboxylat
Methyl-5-fluor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇FO₂S Molecular Weight: 210.225 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371535 CAS-Nummer: 154630-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...