Methyl-7-fluor-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
PC8596-1G
PC8596-5G
APOSPC8596-1GEA
74.1
EUR
APOSPC8596-1G
APOSPC8596-5G
Methyl-7-fluor-2-benzo[b]thiophencarboxylat
Methyl-7-fluor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₇FO₂S Schmelzpunkt: 84…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027102 CAS-Nummer: 550998-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...