4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
PC8615-1G
PC8615-5G
APOSPC8615-1GEA
40.6
EUR
APOSPC8615-1G
APOSPC8615-5G
4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
4-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Formel:
C₁₀H₅F₃O₂S Molecular Weight: 246.21 g/mol Schmelzpunkt: 188…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027109 CAS-Nummer: 826995-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...