Ethyl-2-(3-chloro-4-fluoroanilino)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl [(3-chloro-4-fluorophenyl)amino]acetate

PC8623-1G PC8623-250MG PC8623-500MG PC8623-5G
APOSPC8623-1GEA 169 EUR
APOSPC8623-1G APOSPC8623-250MG APOSPC8623-500MG APOSPC8623-5G
Ethyl-2-(3-chloro-4-fluoroanilino)acetate
Ethyl-2-(3-chloro-4-fluoroanilino)acetate
Formel: C₁₀H₁₁ClFNO₂
Molecular Weight: 231,66 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689674
CAS-Nummer: 2344-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR