Methyl-4-amino-3-fluorbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-amino-3-fluorobenzoate

PC8633-25G PC8633-5G
APOSPC8633-25GEA 49.3 EUR
APOSPC8633-25G APOSPC8633-5G
Methyl-4-amino-3-fluorbenzoat
Methyl-4-amino-3-fluorbenzoat
Formel: C₈H₈FNO₂
Molecular Weight: 169,1530 g/mol
Schmelzpunkt: 107…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689703
CAS-Nummer: 185629-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR