1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC8638-1G PC8638-5G PC8638-10G
APOSPC8638-1GEA 336 EUR
APOSPC8638-1G APOSPC8638-5G APOSPC8638-10G
1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Formel: C₉H₁₁F₃N₂O₂
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04124379
CAS-Nummer: 142818-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR