1-Iodo-2-(pentafluoroethyl)cyclohexane 97%
Lieferant: Apollo Scientific
Synonyme:
cis/trans-1-Iodo-2-(pentafluoroethyl)cyclohexane
PC8651-100MG
PC8651-250MG
APOSPC8651-100MGEA
143
EUR
APOSPC8651-100MG
APOSPC8651-250MG
1-Iodo-2-(pentafluoroethyl)cyclohexane 97%
1-Iodo-2-(pentafluoroethyl)cyclohexane
Formel:
C₈H₁₀F₅I Molecular Weight: 328.07 g/mol Siedepunkt: 60…70 °C/6mmHg Dichte: 1.77 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02183509 CAS-Nummer: 711-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...