Fmoc-3-fluoro-L-phenylalanine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Fmoc-m-fluoro-Phe-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-fluorophenyl)propanoic acid, Fmoc-Phe(3-F)-OH
PC8671-1G
PC8671-5G
APOSPC8671-1GEA
26.1
EUR
APOSPC8671-1G
APOSPC8671-5G
Fmoc-3-fluoro-L-phenylalanine 97%
Fmoc-3-fluoro-L-phenylalanine
Formel:
C₂₄H₂₀FNO₄ Molecular Weight: 405.425 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00672328 CAS-Nummer: 198560-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...