3-Nitro-5-(trifluormethyl)anilin
Lieferant: Apollo Scientific
|
Gefahr
|
PC8673-1G
PC8673-25G
PC8673-5G
APOSPC8673-1GEA
21.9
EUR
APOSPC8673-1G
APOSPC8673-25G
APOSPC8673-5G
3-Nitro-5-(trifluormethyl)anilin
3-Nitro-5-(trifluormethyl)anilin
Formel:
C₇H₅F₃N₂O₂ Molecular Weight: 206,12 g/mol Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061229 CAS-Nummer: 401-94-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...