Kalium(brommethyl)trifluoroborat

Lieferant: Apollo Scientific
Achtung

Synonyme: (Brommethyl)trifluorborsäure Kaliumsalz

PC8705-1G PC8705-25G PC8705-5G
APOSPC8705-1GEA 14.7 EUR
APOSPC8705-1G APOSPC8705-25G APOSPC8705-5G
Kalium(brommethyl)trifluoroborat
Kalium(brommethyl)trifluoroborat
Formel: CH₂BBrF₃K
Molecular Weight: 200.83 g/mol
Schmelzpunkt: 225…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09265154
CAS-Nummer: 888711-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR