Fmoc-4-fluoro-L-phenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine

PC8734-1G PC8734-25G PC8734-5G
APOSPC8734-1GEA 62.5 EUR
APOSPC8734-1G APOSPC8734-25G APOSPC8734-5G
Fmoc-4-fluoro-L-phenylalanine 97%
Fmoc-4-fluoro-L-phenylalanine
Formel: C₂₄H₂₀FNO₄
Molecular Weight: 405,425 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00191197
CAS-Nummer: 169243-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR