2-Brom-1-ethylpyridiniumtetrafluoroborat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate
PC8735-25G
PC8735-5G
PC8735-1G
PC8735-100G
APOSPC8735-25GEA
114
EUR
APOSPC8735-25G
APOSPC8735-5G
APOSPC8735-1G
APOSPC8735-100G
2-Brom-1-ethylpyridiniumtetrafluoroborat 98%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formel:
C₇H₉BBrF₄N Molecular Weight: 273,86 g/mol Schmelzpunkt: 100…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060046 CAS-Nummer: 878-23-9 EINECS: 212-900-2 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...