Methyl-2-fluor-4-nitrobenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-fluoro-4-nitrobenzoate

PC8742-10G PC8742-1G PC8742-5G
APOSPC8742-10GEA 34.8 EUR
APOSPC8742-10G APOSPC8742-1G APOSPC8742-5G
Methyl-2-fluor-4-nitrobenzoat
Methyl-2-fluor-4-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,1400 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444028
CAS-Nummer: 392-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR