5-Amino-3-(3-(trifluormethyl)phenyl)isoxazol
Lieferant: Apollo Scientific
|
Achtung
|
PC8744-10G
PC8744-1G
PC8744-5G
APOSPC8744-10GEA
696
EUR
APOSPC8744-10G
APOSPC8744-1G
APOSPC8744-5G
5-Amino-3-(3-(trifluormethyl)phenyl)isoxazol
5-Amino-3-(3-(trifluoromethyl)phenyl)isoxazole
Formel:
C₁₀H₇F₃N₂O Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787059 CAS-Nummer: 82360-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...