2-Fluorisobuttersäure 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC8763-25G
PC8763-5G
PC8763-1G
APOSPC8763-25GEA
277
EUR
APOSPC8763-25G
APOSPC8763-5G
APOSPC8763-1G
2-Fluorisobuttersäure 95%
2-Fluorisobuttersäure
Formel:
C₄H₇FO₂ Molecular Weight: 106,1 g/mol Siedepunkt: 57…58 °C/10mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03452788 CAS-Nummer: 63812-15-7 UN: 2926 ADR: 4.1+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...