3-Fluorisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoropyridine-4-carboxylic acid

PC8767-25G
APOSPC8767-25GEA 63.8 EUR
APOSPC8767-25G
3-Fluorisonicotinsäure
3-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
Schmelzpunkt: 251…254 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03789251
CAS-Nummer: 393-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR