1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine

Lieferant: Apollo Scientific

Synonyme: 4-(2-Fluoro-4-nitrophenyl)piperazine, N1-BOC protected, tert-Butyl 4-(2-fluoro-4-nitrophenyl)piperazine-1-carboxylate

PC8776-10G PC8776-1G PC8776-5G
APOSPC8776-10GEA 253 EUR
APOSPC8776-10G APOSPC8776-1G APOSPC8776-5G
1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine
1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine
Formel: C₁₅H₂₀FN₃O₄
Molecular Weight: 325.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08443959
CAS-Nummer: 154590-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR